Anti-TFR2 Mouse Monoclonal Antibody [clone: 1D8]

Lieferant: OriGene
ORIGCF504590EA 749 EUR
ORIGCF504590
Anti-TFR2 Mouse Monoclonal Antibody [clone: 1D8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TFR2
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR