Anti-TPD52L3 Mouse Monoclonal Antibody [clone: 4G6]

Lieferant: OriGene
ORIGCF504966EA 749 EUR
ORIGCF504966
Anti-TPD52L3 Mouse Monoclonal Antibody [clone: 4G6]
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TPD52L3
Clonality: Monoclonal
Clone: 4G6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR