Anti-PDIA3 Mouse Monoclonal Antibody [clone: 4D7]

Lieferant: OriGene
ORIGCF505008EA 749 EUR
ORIGCF505008
Anti-PDIA3 Mouse Monoclonal Antibody [clone: 4D7]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: PDIA3
Clonality: Monoclonal
Clone: 4D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR