Anti-MDM4 Mouse Monoclonal Antibody [clone: 1E8]

Lieferant: OriGene
ORIGCF505732EA 749 EUR
ORIGCF505732
Anti-MDM4 Mouse Monoclonal Antibody [clone: 1E8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MDM4
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR