Anti-BBOX1 Mouse Monoclonal Antibody [clone: 4F9]

Lieferant: OriGene
ORIGCF505753EA 749 EUR
ORIGCF505753
Anti-BBOX1 Mouse Monoclonal Antibody [clone: 4F9]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: BBOX1
Clonality: Monoclonal
Clone: 4F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR