Anti-CRYZL1 Mouse Monoclonal Antibody [clone: 1C9]

Lieferant: OriGene
ORIGCF504607EA 749 EUR
ORIGCF504607
Anti-CRYZL1 Mouse Monoclonal Antibody [clone: 1C9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CRYZL1
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR