Anti-C17orf37 Mouse Monoclonal Antibody [clone: 1G2]
Lieferant: OriGene
ORIGCF504614EA 749 EUR
ORIGCF504614
Anti-C17orf37 Mouse Monoclonal Antibody [clone: 1G2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: C17orf37
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human