Anti-TMEM173 Mouse Monoclonal Antibody [clone: 1A3]

Lieferant: OriGene
ORIGCF505016EA 749 EUR
ORIGCF505016
Anti-TMEM173 Mouse Monoclonal Antibody [clone: 1A3]
Antikörper
Recommended Dilutions: Western Blot: 1:200-1:1000

Type: Primary
Antigen: TMEM173
Clonality: Monoclonal
Clone: 1A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR