Anti-TMEM173 Mouse Monoclonal Antibody [clone: 2C8]

Lieferant: OriGene
ORIGCF505020EA 749 EUR
ORIGCF505020
Anti-TMEM173 Mouse Monoclonal Antibody [clone: 2C8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TMEM173
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR