Anti-TMEM173 Mouse Monoclonal Antibody [clone: 3B10]

Lieferant: OriGene
ORIGCF505021EA 749 EUR
ORIGCF505021
Anti-TMEM173 Mouse Monoclonal Antibody [clone: 3B10]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TMEM173
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR