Anti-ZBED1 Mouse Monoclonal Antibody [clone: 2H3]

Lieferant: OriGene
ORIGCF505045EA 749 EUR
ORIGCF505045
Anti-ZBED1 Mouse Monoclonal Antibody [clone: 2H3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ZBED1
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR