Anti-SYT4 Mouse Monoclonal Antibody [clone: 3D4]

Lieferant: OriGene
ORIGCF505860EA 770 EUR
ORIGCF505860
Anti-SYT4 Mouse Monoclonal Antibody [clone: 3D4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SYT4
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR