Anti-CALR3 Mouse Monoclonal Antibody [clone: 1D1]
Lieferant: OriGene
ORIGCF505873EA 749 EUR
ORIGCF505873
Anti-CALR3 Mouse Monoclonal Antibody [clone: 1D1]
Antikörper
Recommended Dilutions: Western Blot: 1:4000
Type: Primary
Antigen: CALR3
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human