Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 1D12]

Lieferant: OriGene
ORIGCF504671EA 749 EUR
ORIGCF504671
Anti-UBE2E3 Mouse Monoclonal Antibody [clone: 1D12]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: UBE2E3
Clonality: Monoclonal
Clone: 1D12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR