Anti-TRIM38 Mouse Monoclonal Antibody [clone: 1B9]

Lieferant: OriGene
ORIGCF505962EA 749 EUR
ORIGCF505962
Anti-TRIM38 Mouse Monoclonal Antibody [clone: 1B9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TRIM38
Clonality: Monoclonal
Clone: 1B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR