Anti-TRIM38 Mouse Monoclonal Antibody [clone: 2G9]

Lieferant: OriGene
ORIGCF505969EA 749 EUR
ORIGCF505969
Anti-TRIM38 Mouse Monoclonal Antibody [clone: 2G9]
Antikörper
Recommended Dilutions: Western Blot: 1:200; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: TRIM38
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR