Anti-CTAG1B Mouse Monoclonal Antibody [clone: 5E1]

Lieferant: OriGene
ORIGCF505972EA 749 EUR
ORIGCF505972
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 5E1]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CTAG1B
Clonality: Monoclonal
Clone: 5E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR