Anti-PDSS2 Mouse Monoclonal Antibody [clone: 1D2]

Lieferant: OriGene
ORIGCF503935EA 749 EUR
ORIGCF503935
Anti-PDSS2 Mouse Monoclonal Antibody [clone: 1D2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PDSS2
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR