Anti-RFPL3 Mouse Monoclonal Antibody [clone: 2B6]

Lieferant: OriGene
ORIGCF504732EA 770 EUR
ORIGCF504732
Anti-RFPL3 Mouse Monoclonal Antibody [clone: 2B6]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: RFPL3
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR