Anti-SELENBP1 Mouse Monoclonal Antibody [clone: 2B12]

Lieferant: OriGene
ORIGCF504740EA 749 EUR
ORIGCF504740
Anti-SELENBP1 Mouse Monoclonal Antibody [clone: 2B12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: SELENBP1
Clonality: Monoclonal
Clone: 2B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR