Anti-CPO Mouse Monoclonal Antibody [clone: 1G3]
Lieferant: OriGene
ORIGCF504750EA 749 EUR
ORIGCF504750
Anti-CPO Mouse Monoclonal Antibody [clone: 1G3]
Antikörper
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: CPO
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human