Anti-PHF7 Mouse Monoclonal Antibody [clone: 1F7]
Lieferant: OriGene
ORIGCF505144EA 749 EUR
ORIGCF505144
Anti-PHF7 Mouse Monoclonal Antibody [clone: 1F7]
Antikörper
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: PHF7
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human