Anti-MAPK13 Mouse Monoclonal Antibody [clone: 12B12]

Lieferant: OriGene
ORIGCF505781EA 749 EUR
ORIGCF505781
Anti-MAPK13 Mouse Monoclonal Antibody [clone: 12B12]
Antikörper
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK13
Clonality: Monoclonal
Clone: 12B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR