Anti-BMP1 Mouse Monoclonal Antibody [clone: 3E9]

Lieferant: OriGene
ORIGCF505813EA 749 EUR
ORIGCF505813
Anti-BMP1 Mouse Monoclonal Antibody [clone: 3E9]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: BMP1
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR