Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 2A6]

Lieferant: OriGene
ORIGCF505816EA 749 EUR
ORIGCF505816
Anti-PIK3R5 Mouse Monoclonal Antibody [clone: 2A6]
Antikörper
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PIK3R5
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR