Anti-ERBB2 Mouse Monoclonal Antibody [clone: 6H9]

Lieferant: OriGene
ORIGCF505829EA 749 EUR
ORIGCF505829
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 6H9]
Antikörper
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 6H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR