Anti-SERPINB2 Mouse Monoclonal Antibody [clone: 4A3]

Lieferant: OriGene
ORIGCF504003EA 749 EUR
ORIGCF504003
Anti-SERPINB2 Mouse Monoclonal Antibody [clone: 4A3]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINB2
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR