Anti-DOK7 Mouse Monoclonal Antibody [clone: 4C6]

Lieferant: OriGene
ORIGCF504786EA 749 EUR
ORIGCF504786
Anti-DOK7 Mouse Monoclonal Antibody [clone: 4C6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: DOK7
Clonality: Monoclonal
Clone: 4C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR