Anti-PHF7 Mouse Monoclonal Antibody [clone: 1A11]

Lieferant: OriGene
ORIGCF505154EA 749 EUR
ORIGCF505154
Anti-PHF7 Mouse Monoclonal Antibody [clone: 1A11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PHF7
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR