Anti-STK38L Mouse Monoclonal Antibody [clone: 4D8]
Lieferant: OriGene
ORIGCF505176EA 770 EUR
ORIGCF505176
Anti-STK38L Mouse Monoclonal Antibody [clone: 4D8]
Antikörper
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100
Type: Primary
Antigen: STK38L
Clonality: Monoclonal
Clone: 4D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat