Anti-COPS6 Mouse Monoclonal Antibody [clone: 4E7]

Lieferant: OriGene
ORIGCF505897EA 749 EUR
ORIGCF505897
Anti-COPS6 Mouse Monoclonal Antibody [clone: 4E7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: COPS6
Clonality: Monoclonal
Clone: 4E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR