Anti-ENPEP Mouse Monoclonal Antibody [clone: 1D4]

Lieferant: OriGene
ORIGCF504037EA 749 EUR
ORIGCF504037
Anti-ENPEP Mouse Monoclonal Antibody [clone: 1D4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ENPEP
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR