Anti-PPIL6 Mouse Monoclonal Antibody [clone: 2G3]

Lieferant: OriGene
ORIGCF504840EA 749 EUR
ORIGCF504840
Anti-PPIL6 Mouse Monoclonal Antibody [clone: 2G3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PPIL6
Clonality: Monoclonal
Clone: 2G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR