Anti-MRPS34 Mouse Monoclonal Antibody [clone: 4C4]

Lieferant: OriGene
ORIGCF505232EA 749 EUR
ORIGCF505232
Anti-MRPS34 Mouse Monoclonal Antibody [clone: 4C4]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: MRPS34
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR