Anti-C4orf22 Mouse Monoclonal Antibody [clone: 1A6]

Lieferant: OriGene
ORIGCF505987EA 770 EUR
ORIGCF505987
Anti-C4orf22 Mouse Monoclonal Antibody [clone: 1A6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: C4orf22
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR