Anti-IL1F9 Mouse Monoclonal Antibody [clone: 6E4]

Lieferant: OriGene
ORIGCF505995EA 749 EUR
ORIGCF505995
Anti-IL1F9 Mouse Monoclonal Antibody [clone: 6E4]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: IL1F9
Clonality: Monoclonal
Clone: 6E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR