Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 1B4]

Lieferant: OriGene
ORIGCF506010EA 749 EUR
ORIGCF506010
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 1B4]
Antikörper
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR