Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 9E9]

Lieferant: OriGene
ORIGCF506033EA 749 EUR
ORIGCF506033
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 9E9]
Antikörper
Recommended Dilutions: Western Blot: 1:8000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 9E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR