Anti-PSMA4 Mouse Monoclonal Antibody [clone: 4B8]

Lieferant: OriGene
ORIGCF504059EA 749 EUR
ORIGCF504059
Anti-PSMA4 Mouse Monoclonal Antibody [clone: 4B8]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: PSMA4
Clonality: Monoclonal
Clone: 4B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR