Anti-ETF1 Mouse Monoclonal Antibody [clone: 2E3]

Lieferant: OriGene
ORIGCF502891EA 749 EUR
ORIGCF502891
Anti-ETF1 Mouse Monoclonal Antibody [clone: 2E3]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: ETF1
Clonality: Monoclonal
Clone: 2E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR