Anti-C1orf50 Mouse Monoclonal Antibody [clone: 1B11]

Lieferant: OriGene
ORIGCF502912EA 770 EUR
ORIGCF502912
Anti-C1orf50 Mouse Monoclonal Antibody [clone: 1B11]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C1orf50
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR