Anti-PPIL3 Mouse Monoclonal Antibody [clone: 1G3]
Lieferant: OriGene
ORIGCF505245EA 749 EUR
ORIGCF505245
Anti-PPIL3 Mouse Monoclonal Antibody [clone: 1G3]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100
Type: Primary
Antigen: PPIL3
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human