Anti-FATE1 Mouse Monoclonal Antibody [clone: 7B12]

Lieferant: OriGene
ORIGCF505250EA 749 EUR
ORIGCF505250
Anti-FATE1 Mouse Monoclonal Antibody [clone: 7B12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: FATE1
Clonality: Monoclonal
Clone: 7B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR