Anti-UBE2G2 Mouse Monoclonal Antibody [clone: 9F2]

Lieferant: OriGene
ORIGCF505288EA 770 EUR
ORIGCF505288
Anti-UBE2G2 Mouse Monoclonal Antibody [clone: 9F2]
Antikörper
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: UBE2G2
Clonality: Monoclonal
Clone: 9F2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR