Anti-ERCC4 Mouse Monoclonal Antibody [clone: 3H10]

Lieferant: OriGene
ORIGCF503324EA 749 EUR
ORIGCF503324
Anti-ERCC4 Mouse Monoclonal Antibody [clone: 3H10]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC4
Clonality: Monoclonal
Clone: 3H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR