Anti-NEU2 Mouse Monoclonal Antibody [clone: 8C12]

Lieferant: OriGene
ORIGCF503332EA 770 EUR
ORIGCF503332
Anti-NEU2 Mouse Monoclonal Antibody [clone: 8C12]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NEU2
Clonality: Monoclonal
Clone: 8C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR