Anti-DOK2 Mouse Monoclonal Antibody [clone: 3D6]

Lieferant: OriGene
ORIGCF503337EA 749 EUR
ORIGCF503337
Anti-DOK2 Mouse Monoclonal Antibody [clone: 3D6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DOK2
Clonality: Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR