Anti-KCNJ3 Mouse Monoclonal Antibody [clone: 2F2]

Lieferant: OriGene
ORIGCF504137EA 749 EUR
ORIGCF504137
Anti-KCNJ3 Mouse Monoclonal Antibody [clone: 2F2]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: KCNJ3
Clonality: Monoclonal
Clone: 2F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR