Anti-STAT4 Mouse Monoclonal Antibody [clone: 4H6]

Lieferant: OriGene
ORIGCF502940EA 770 EUR
ORIGCF502940
Anti-STAT4 Mouse Monoclonal Antibody [clone: 4H6]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT4
Clonality: Monoclonal
Clone: 4H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR