Anti-PBX1 Mouse Monoclonal Antibody [clone: 1H5]

Lieferant: OriGene
ORIGCF502941EA 749 EUR
ORIGCF502941
Anti-PBX1 Mouse Monoclonal Antibody [clone: 1H5]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PBX1
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR